siRNA Details
| Gene Symbol: MRPL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014175.3 | NP_054894.1 | 4 | 511 | ||
| XM_017013362.1 | XP_016868851.1 | 4 | 522 | |||
| GenBank mRNA | AF161494.1 | - | 4 | 503 | ||
| AK312832.1 | - | 4 | 475 | |||
| AL136665.1 | - | 4 | 481 | |||
| BC000891.1 | - | 4 | 511 | 3463430 | ||
| CR533531.1 | - | 4 | 448 | |||

MRPL15
Genomic Map