siRNA Details
| Gene Symbol: PATZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014323.2 | NP_055138.2 | 2 | 1956 | ||
| NM_032050.1 | NP_114439.1 | 2 | 1956 | |||
| NM_032051.1 | NP_114440.1 | 2 | 1956 | |||
| NM_032052.1 | NP_114441.1 | 2 | 1956 | |||
| GenBank mRNA | AB208787.1 | - | 3 | 1326 | ||
| AF119256.1 | - | 2 | 1955 | |||
| AF254082.1 | - | 2 | 1925 | |||
| AF254083.1 | - | 2 | 1925 | |||
| AF254084.1 | - | 2 | 1925 | |||
| AF254085.1 | - | 2 | 1925 | |||
| AK123441.2 | - | 1 | 655 | |||
| AK291803.1 | - | 2 | 1469 | |||
| AL096880.1 | - | 2 | 1290 | |||
| AY028384.1 | - | 2 | 1686 | |||
| BC021091.1 | - | 2 | 1503 | 4857537 | ||
| CR456613.1 | - | 2 | 1394 | |||

PATZ1
Genomic Map