siRNA Details
| Gene Symbol: DAPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014326.3 | NP_055141.2 | 9 | 856 | ||
| XM_011521413.2 | XP_011519715.1 | 8 | 831 | |||
| XM_011521414.1 | XP_011519716.1 | 8 | 831 | |||
| XM_011521415.2 | XP_011519717.1 | 8 | 831 | |||
| XM_011521416.2 | XP_011519718.1 | 9 | 1004 | |||
| XM_011521421.2 | XP_011519723.1 | 7 | 641 | |||
| XM_017022043.1 | XP_016877532.1 | 7 | 600 | |||
| XM_017022044.1 | XP_016877533.1 | 8 | 831 | |||
| XM_017022047.1 | XP_016877536.1 | 7 | 641 | |||
| XM_017022048.1 | XP_016877537.1 | 6 | 590 | |||
| XM_017022049.1 | XP_016877538.1 | 7 | 637 | |||
| XM_017022050.1 | XP_016877539.1 | 6 | 442 | |||
| XM_017022051.1 | XP_016877540.1 | 6 | 588 | |||
| XM_017022052.1 | XP_016877541.1 | 3 | 1124 | |||
| GenBank mRNA | AB018001.1 | - | 9 | 855 | ||
| AF052941.1 | - | 9 | 856 | |||
| BC114506.1 | - | 8 | 845 | 40005390 | ||
| BC114951.1 | - | 8 | 753 | 40005391 | ||
| GU056176.1 | - | 9 | 856 | |||

DAPK2
Genomic Map