siRNA Details
| Gene Symbol: ZNF318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014345.2 | NP_055160.2 | 3 | 1076 | ||
| XM_011514451.2 | XP_011512753.1 | 3 | 677 | |||
| XM_011514452.2 | XP_011512754.1 | 3 | 1309 | |||
| GenBank mRNA | AF090114.1 | - | 3 | 1079 | ||
| AF121141.1 | - | 3 | 814 | |||
| BC030687.1 | - | 3 | 1111 | |||
| BC098434.1 | - | 3 | 1051 | |||

ZNF318
Genomic Map