siRNA Details
| Gene Symbol: ZNF318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014345.2 | NP_055160.2 | 4 | 2545 | ||
| XM_011514451.2 | XP_011512753.1 | 4 | 2146 | |||
| XM_011514452.2 | XP_011512754.1 | 4 | 2778 | |||
| GenBank mRNA | AF090114.1 | - | 4 | 2548 | ||
| AF121141.1 | - | 4 | 2283 | |||
| BC030687.1 | - | 4 | 2580 | |||
| BC098434.1 | - | 4 | 2520 | |||

ZNF318
Genomic Map