siRNA Details
| Gene Symbol: SACS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278055.1 | NP_001264984.1 | 8 | 6062 | ||
| NM_014363.5 | NP_055178.3 | 10 | 6593 | |||
| XM_005266338.2 | XP_005266395.1 | 11 | 6620 | |||
| XM_011535039.2 | XP_011533341.1 | 10 | 6228 | |||
| XM_017020539.1 | XP_016876028.1 | 9 | 6201 | |||
| GenBank mRNA | CR749427.1 | - | 3 | 4830 | ||

SACS
Genomic Map