siRNA Details
| Gene Symbol: RNF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014372.4 | NP_055187.1 | 3 | 1684 | ||
| GenBank mRNA | AB024703.1 | - | 3 | 1326 | ||
| AF151881.1 | - | 3 | 1327 | |||
| AL049294.1 | - | 1 | 464 | |||
| BC020964.2 | - | 3 | 1279 | 3917224 | ||
| BC047654.1 | - | 3 | 1651 | 5261966 | ||

RNF11
Genomic Map