siRNA Details
| Gene Symbol: GPR160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014373.2 | NP_055188.1 | 4 | 1830 | ||
| XM_005247346.3 | XP_005247403.1 | 5 | 2440 | |||
| XM_005247347.3 | XP_005247404.1 | 3 | 1476 | |||
| XM_006713582.3 | XP_006713645.1 | 4 | 1792 | |||
| XM_011512678.2 | XP_011510980.1 | 5 | 2122 | |||
| XM_017006161.1 | XP_016861650.1 | 5 | 1933 | |||
| XM_017006162.1 | XP_016861651.1 | 5 | 1937 | |||
| XM_017006163.1 | XP_016861652.1 | 4 | 2454 | |||
| GenBank mRNA | AJ249248.1 | - | 3 | 1558 | ||

GPR160
Genomic Map