siRNA Details
| Gene Symbol: LAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014987.1 | NP_001014987.1 | 7 | 732 | ||
| NM_001014988.1 | NP_001014988.1 | 7 | 729 | |||
| NM_001014989.1 | NP_001014989.2 | 8 | 524 | |||
| NM_014387.3 | NP_055202.1 | 6 | 819 | |||
| GenBank mRNA | AF036905.1 | - | 7 | 437 | ||
| AF036906.1 | - | 6 | 545 | |||
| AJ223280.1 | - | 7 | 702 | |||
| BC011563.1 | - | 8 | 524 | 4644259 | ||
| BI911769.1 | - | 7 | 698 | |||
| FN432831.1 | - | 6 | 494 | |||
| FN432832.1 | - | 6 | 464 | |||
| FN432833.1 | - | 5 | 437 | |||
| FN432834.1 | - | 6 | 351 | |||

LAT
Genomic Map