siRNA Details
| Gene Symbol: GHITM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014394.2 | NP_055209.2 | 4 | 495 | ||
| GenBank mRNA | AB009685.1 | - | 3 | 291 | ||
| AF060923.1 | - | 4 | 369 | |||
| AF078863.1 | - | 4 | 420 | |||
| AF131820.1 | - | 4 | 422 | |||
| AF161400.1 | - | 4 | 404 | |||
| AK130094.1 | - | 4 | 398 | |||
| AK130097.1 | - | 4 | 363 | |||
| AK225318.1 | - | 4 | 372 | |||
| AK292864.1 | - | 4 | 363 | |||
| AK297960.1 | - | 4 | 362 | |||
| AK298332.1 | - | 4 | 377 | |||
| AK299937.1 | - | 3 | 401 | |||
| AL136713.1 | - | 4 | 405 | |||
| AY358824.1 | - | 4 | 370 | |||
| BC010354.1 | - | 4 | 366 | 4093037 | ||
| BC022872.1 | - | 4 | 377 | |||
| CR533519.1 | - | 3 | 291 | |||

GHITM
Genomic Map