siRNA Details
| Gene Symbol: SNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003498.5 | NP_003489.1 | 2 | 2835 | ||
| XM_017023741.1 | XP_016879230.1 | 2 | 2863 | |||
| XM_017023742.1 | XP_016879231.1 | 2 | 2761 | |||
| GenBank mRNA | AF070673.1 | - | 2 | 2818 | ||
| AK025795.1 | - | 1 | 1907 | |||
| AL161976.1 | - | 2 | 2798 | |||
| BC036100.1 | - | 2 | 2813 | 5277179 | ||
| BC036443.1 | - | 2 | 2807 | 4823944 | ||

SNN
Genomic Map