siRNA Details
| Gene Symbol: INVS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318381.1 | NP_001305310.1 | 18 | 3939 | ||
| NM_001318382.1 | NP_001305311.1 | 17 | 3862 | |||
| NM_014425.4 | NP_055240.2 | 17 | 3851 | |||
| NR_134606.1 | 17 | 3802 | ||||
| GenBank mRNA | AI792224.1 | - | 1 | 376 | ||
| BC111761.1 | - | 16 | 3634 | 40012263 | ||
| BC112351.1 | - | 16 | 3585 | 40012265 | ||

INVS
Genomic Map