siRNA Details
| Gene Symbol: ZNF330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001292002.1 | NP_001278931.1 | 8 | 828 | ||
| NM_014487.5 | NP_055302.1 | 9 | 899 | |||
| XM_017008033.1 | XP_016863522.1 | 9 | 1043 | |||
| GenBank mRNA | AJ006591.2 | - | 9 | 844 | ||
| AK002106.1 | - | 9 | 848 | |||
| AK223609.1 | - | 9 | 850 | |||
| AK294089.1 | - | 8 | 782 | |||
| AK315463.1 | - | 9 | 885 | |||
| BC004421.1 | - | 9 | 875 | 3352916 | ||

ZNF330
Genomic Map