siRNA Details
| Gene Symbol: GOLIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014498.4 | NP_055313.1 | 7 | 1305 | ||
| XM_005247364.2 | XP_005247421.1 | 7 | 1304 | |||
| GenBank mRNA | AK307557.1 | - | 7 | 1010 | ||
| AK307718.1 | - | 7 | 980 | |||
| U55853.1 | - | 7 | 678 | |||

GOLIM4
Genomic Map