siRNA Details
| Gene Symbol: SLC25A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165417.1 | NP_001158889.1 | 8 | 1304 | ||
| NM_001165418.1 | NP_001158890.1 | 7 | 1184 | |||
| NM_003562.4 | NP_003553.2 | 8 | 1337 | |||
| GenBank mRNA | AF070548.1 | - | 8 | 1191 | ||
| AK290376.1 | - | 8 | 1088 | |||
| BC006508.2 | - | 8 | 1193 | 2960933 | ||
| BC006519.2 | - | 8 | 1193 | 2960933 | ||
| BC016294.2 | - | 8 | 1112 | 3911858 | ||
| BC017170.2 | - | 8 | 1193 | 2960933 | ||

SLC25A11
Genomic Map