siRNA Details
| Gene Symbol: ZIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146326.1 | NP_001139798.1 | 12 | 1471 | ||
| NM_001146327.1 | NP_001139799.1 | 12 | 1558 | |||
| NM_015363.4 | NP_056178.3 | 11 | 1395 | |||
| GenBank mRNA | AF166122.1 | - | 11 | 1378 | ||
| BC104876.1 | - | 11 | 1302 | 8143879 | ||
| BC104878.1 | - | 11 | 1386 | 8143881 | ||

ZIM2
Genomic Map