siRNA Details
| Gene Symbol: DCAF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015397.3 | NP_056212.1 | 2, 3 | 682 | ||
| XM_011517843.1 | XP_011516145.1 | 2, 3 | 509 | |||
| GenBank mRNA | AB067479.1 | - | 2, 3 | 602 | ||
| AK000354.1 | - | 2, 3 | 460 | |||
| AK292935.1 | - | 2, 3 | 488 | |||
| BC008893.2 | - | 1, 2 | 245 | |||
| BC063823.1 | - | 2, 3 | 600 | 6184796 | ||
| BG718613.1 | - | 2, 3 | 653 | |||
| DQ518909.1 | - | 2, 3 | 602 | |||

DCAF12
Genomic Map