siRNA Details
| Gene Symbol: RNF19A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001280539.1 | NP_001267468.1 | 4 | 1108 | ||
| NM_015435.4 | NP_056250.3 | 4 | 1139 | |||
| NM_183419.3 | NP_904355.1 | 3 | 1215 | |||
| XM_005250853.3 | XP_005250910.1 | 3 | 1177 | |||
| XM_011516956.2 | XP_011515258.1 | 3 | 1200 | |||
| XM_017013302.1 | XP_016868791.1 | 4 | 1300 | |||
| XM_017013303.1 | XP_016868792.1 | 4 | 1210 | |||
| XM_017013304.1 | XP_016868793.1 | 3 | 1157 | |||
| GenBank mRNA | AB029316.1 | - | 4 | 1139 | ||
| BC093938.1 | - | 2 | 911 | 7939783 | ||
| BC093940.2 | - | 2 | 911 | 7939785 | ||

RNF19A
Genomic Map