siRNA Details
| Gene Symbol: SEC31B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015490.3 | NP_056305.1 | 16 | 2073 | ||
| GenBank mRNA | AF274863.1 | - | 16 | 2069 | ||
| AK128343.1 | - | 10 | 2207 | |||
| AL080141.1 | - | 9 | 1181 | |||
| BC034946.1 | - | 16 | 2629 | |||
| BC044569.1 | - | 16 | 2622 | |||
| BC053949.1 | - | 16 | 2622 | |||
| BC071580.1 | - | 16 | 2549 | |||
| BC144313.1 | - | 15 | 2011 | 9052833 | ||
| BX640914.1 | - | 16 | 1970 | |||

SEC31B
Genomic Map