siRNA Details
| Gene Symbol: TIPARP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184717.1 | NP_001171646.1 | 2 | 1279 | ||
| NM_001184718.1 | NP_001171647.1 | 2 | 844 | |||
| NM_015508.4 | NP_056323.2 | 2 | 1194 | |||
| GenBank mRNA | AK097785.1 | - | 2 | 1023 | ||
| AK291261.1 | - | 2 | 1029 | |||
| BC034397.1 | - | 2 | 844 | |||
| BC050350.1 | - | 2 | 1001 | 5264807 | ||
| BX537965.1 | - | 2 | 1021 | |||
| CR749647.1 | - | 2 | 1280 | |||

TIPARP
Genomic Map