siRNA Details
Gene Symbol: UPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015542.3 | NP_056357.1 | 3 | 539 | ||
NM_080599.2 | NP_542166.1 | 3 | 709 | |||
XM_011519449.2 | XP_011517751.1 | 3 | 587 | |||
XM_017016034.1 | XP_016871523.1 | 2 | 85 | |||
GenBank mRNA | AB037829.1 | - | 2 | 908 | ||
AF301013.1 | - | 3 | 532 | |||
AF318574.1 | - | 3 | 509 | |||
AK096191.1 | - | 4 | 449 | |||
AK310925.1 | - | 3 | 498 | |||
AY013249.1 | - | 2 | 458 | |||
BC114964.1 | - | 3 | 503 | 40082739 | ||
BC115737.1 | - | 3 | 503 | 40082728 | ||
DA188353.1 | - | 3 | 539 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
