siRNA Details
| Gene Symbol: LTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320766.1 | NP_001307695.1 | 7 | 1014 | ||
| NM_015565.2 | NP_056380.2 | 7 | 1011 | |||
| XM_006723987.3 | XP_006724050.1 | 7 | 1009 | |||
| XM_017028316.1 | XP_016883805.1 | 6 | 691 | |||
| GenBank mRNA | AB018257.2 | - | 7 | 902 | ||
| AF161350.1 | - | 7 | 898 | |||
| AK001915.1 | - | 7 | 958 | |||
| AK023499.1 | - | 6 | 645 | |||
| BC029371.1 | - | 7 | 918 | |||
| BC140790.1 | - | 7 | 924 | 9021658 | ||
| BC150284.1 | - | 7 | 902 | 8860138 | ||

LTN1
Genomic Map