siRNA Details
| Gene Symbol: GPKOW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015698.5 | NP_056513.2 | 4 | 552 | ||
| XM_017029415.1 | XP_016884904.1 | 5 | 626 | |||
| GenBank mRNA | AB209806.1 | - | 4 | 472 | ||
| BC000397.2 | - | 4 | 480 | 2822421 | ||
| BC003148.1 | - | 4 | 480 | 2822421 | ||
| BE795098.1 | - | 552 | ||||
| U66359.1 | - | 4 | 530 | |||

GPKOW
Genomic Map