siRNA Details
| Gene Symbol: PLA2G3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015715.4 | NP_056530.2 | 7 | 2225 | ||
| XM_011530204.1 | XP_011528506.1 | 7 | 2447 | |||
| XM_011530205.1 | XP_011528507.1 | 7 | 2436 | |||
| GenBank mRNA | AF220490.1 | - | 7 | 2225 | ||
| BC025316.1 | - | 7 | 2101 | 5458919 | ||

PLA2G3
Genomic Map