siRNA Details
| Gene Symbol: CUTC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015960.2 | NP_057044.2 | 6 | 658 | ||
| GenBank mRNA | AF132966.1 | - | 6 | 612 | ||
| AK001142.1 | - | 6 | 578 | |||
| AK302507.1 | - | 6 | 594 | |||
| AK314687.1 | - | 6 | 594 | |||
| BC015059.1 | - | 6 | 554 | |||
| BC021105.1 | - | 6 | 537 | 4547900 | ||
| BC028948.1 | - | 6 | 626 | 4776344 | ||
| CD107559.1 | - | 684 | ||||

CUTC
Genomic Map