siRNA Details
| Gene Symbol: FCF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318508.1 | NP_001305437.1 | 6 | 470 | ||
| NM_015962.4 | NP_057046.1 | 6 | 434 | |||
| XM_005267730.3 | XP_005267787.1 | 6 | 566 | |||
| XM_011536815.2 | XP_011535117.1 | 5 | 391 | |||
| XM_011536816.2 | XP_011535118.1 | 5 | 391 | |||
| GenBank mRNA | AF132969.1 | - | 5 | 383 | ||
| AK290666.1 | - | 6 | 415 | |||
| BC080600.1 | - | 6 | 394 | 4889846 | ||
| BX248271.1 | - | 6 | 396 | |||
| BX489015.1 | - | 6 | 436 | |||
| BX647810.1 | - | 6 | 460 | |||

FCF1
Genomic Map