siRNA Details
| Gene Symbol: HDDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016063.2 | NP_057147.2 | 6 | 1563 | ||
| GenBank mRNA | AF151888.1 | - | 5 | 1320 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

HDDC2| Gene Symbol: HDDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016063.2 | NP_057147.2 | 6 | 1563 | ||
| GenBank mRNA | AF151888.1 | - | 5 | 1320 | ||