siRNA Details
| Gene Symbol: EIF3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242923.1 | NP_001229852.1 | 3 | 217 | ||
| NM_016091.3 | NP_057175.1 | 3 | 217 | |||
| XM_005261625.1 | XP_005261682.1 | 3 | 172 | |||
| XM_006724260.3 | XP_006724323.1 | 3 | 815 | |||
| XM_017028814.1 | XP_016884303.1 | 3 | 172 | |||
| GenBank mRNA | AF077207.1 | - | 3 | 153 | ||
| AF083243.1 | - | 3 | 172 | |||
| AF109359.1 | - | 3 | 154 | |||
| AK056129.1 | - | 3 | 721 | |||
| AK093540.1 | - | 2 | 641 | |||
| AK129648.1 | - | 3 | 170 | |||
| AK222496.1 | - | 3 | 172 | |||
| AK222529.1 | - | 3 | 170 | |||
| AK293629.1 | - | 3 | 172 | |||
| AK298776.1 | - | 3 | 148 | |||
| AK301323.1 | - | 3 | 171 | |||
| AK302346.1 | - | 3 | 172 | |||
| AK315533.1 | - | 3 | 178 | |||
| AL357455.1 | - | 3 | 145 | |||
| BC001101.2 | - | 3 | 155 | 3507964 | ||
| BC007510.2 | - | 3 | 155 | 2959747 | ||
| BC029265.1 | - | 3 | 157 | 5110941 | ||
| CR456436.1 | - | 3 | 278 | |||
| DA051005.1 | - | 3 | 217 | |||
| DA663575.1 | - | 3 | 178 | |||

EIF3L
Genomic Map