siRNA Details
| Gene Symbol: AIG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286587.1 | NP_001273516.1 | 7 | 774 | ||
| NM_016108.3 | NP_057192.2 | 6 | 831 | |||
| XM_011535883.2 | XP_011534185.1 | 7 | 881 | |||
| XM_011535884.2 | XP_011534186.1 | 6 | 2475 | |||
| XM_011535885.2 | XP_011534187.1 | 5 | 696 | |||
| GenBank mRNA | AF151861.1 | - | 7 | 781 | ||
| AF153605.1 | - | 6 | 774 | |||
| AJ420480.1 | - | 4 | 447 | |||
| AK222475.1 | - | 6 | 793 | |||
| BC025278.1 | - | 6 | 762 | 4859216 | ||
| BG196820.1 | - | 1 | 73 | |||
| BX538067.1 | - | 5 | 534 | |||

AIG1
Genomic Map