siRNA Details
| Gene Symbol: KCTD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319294.1 | NP_001306223.1 | 9 | 1006 | ||
| NM_001319295.1 | NP_001306224.1 | 9 | 1078 | |||
| NM_016121.4 | NP_057205.2 | 9 | 1006 | |||
| XM_005273158.1 | XP_005273215.1 | 9 | 1272 | |||
| GenBank mRNA | AK055925.1 | - | 3 | 283 | ||
| AK296275.1 | - | 7 | 539 | |||
| AK316272.1 | - | 5 | 407 | |||
| AL834389.1 | - | 3 | 230 | |||
| BC013868.2 | - | 9 | 993 | |||
| BC094879.1 | - | 9 | 955 | 3078971 | ||
| BC117188.1 | - | 9 | 888 | 40125739 | ||
| BC126173.1 | - | 9 | 888 | 8991889 | ||

KCTD3
Genomic Map