siRNA Details
| Gene Symbol: INSIG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321329.1 | NP_001308258.1 | 6 | 1550 | ||
| NM_001321330.1 | NP_001308259.1 | 5 | 1175 | |||
| NM_001321331.1 | NP_001308260.1 | 6 | 1183 | |||
| NM_001321332.1 | NP_001308261.1 | 6 | 1176 | |||
| NM_001321333.1 | NP_001308262.1 | 5 | 1083 | |||
| NM_016133.3 | NP_057217.2 | 6 | 1557 | |||
| GenBank mRNA | AK021692.1 | - | 5 | 989 | ||
| AL080184.1 | - | 2 | 692 | |||
| BX647805.1 | - | 6 | 1099 | |||

INSIG2
Genomic Map