siRNA Details
| Gene Symbol: INSIG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321329.1 | NP_001308258.1 | 6 | 1596 | ||
| NM_001321330.1 | NP_001308259.1 | 5 | 1221 | |||
| NM_001321331.1 | NP_001308260.1 | 6 | 1229 | |||
| NM_001321332.1 | NP_001308261.1 | 6 | 1222 | |||
| NM_001321333.1 | NP_001308262.1 | 5 | 1129 | |||
| NM_016133.3 | NP_057217.2 | 6 | 1603 | |||
| GenBank mRNA | AF527632.1 | - | 6 | 1552 | ||
| AK021692.1 | - | 5 | 1035 | |||
| AL080184.1 | - | 2 | 738 | |||
| BC022475.1 | - | 6 | 1547 | 4794938 | ||
| BX647805.1 | - | 6 | 1145 | |||

INSIG2
Genomic Map