siRNA Details
| Gene Symbol: CRNN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016190.2 | NP_057274.1 | 3 | 579 | ||
| GenBank mRNA | AF077831.1 | - | 3 | 557 | ||
| AK291301.1 | - | 3 | 568 | |||
| AK298640.1 | - | 4 | 456 | |||
| AK316568.1 | - | 3 | 568 | |||
| BC030807.1 | - | 3 | 568 | 4271624 | ||

CRNN
Genomic Map