siRNA Details
| Gene Symbol: CST7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003650.3 | NP_003641.3 | 3 | 537 | ||
| GenBank mRNA | AF031824.1 | - | 3 | 476 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CST7| Gene Symbol: CST7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003650.3 | NP_003641.3 | 3 | 537 | ||
| GenBank mRNA | AF031824.1 | - | 3 | 476 | ||