siRNA Details
| Gene Symbol: HSD17B11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016245.4 | NP_057329.3 | 7 | 1702 | ||
| GenBank mRNA | AF126780.1 | - | 7 | 1657 | ||
| AF273056.1 | - | 7 | 1555 | |||
| AK075348.1 | - | 7 | 1586 | |||
| AY358553.1 | - | 7 | 1569 | |||
| BC036001.1 | - | 7 | 1593 | 4715042 | ||
| CB163647.1 | - | 1 | 377 | |||

HSD17B11
Genomic Map