siRNA Details
| Gene Symbol: PHF20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016436.4 | NP_057520.2 | 8 | 1192 | ||
| XM_005260421.2 | XP_005260478.1 | 5 | 802 | |||
| XM_011528843.1 | XP_011527145.1 | 7 | 793 | |||
| XM_011528844.1 | XP_011527146.1 | 5 | 864 | |||
| XM_017027864.1 | XP_016883353.1 | 9 | 1274 | |||
| XM_017027865.1 | XP_016883354.1 | 9 | 1475 | |||
| XM_017027866.1 | XP_016883355.1 | 8 | 1395 | |||
| XM_017027867.1 | XP_016883356.1 | 8 | 1230 | |||
| XM_017027868.1 | XP_016883357.1 | 6 | 1016 | |||
| XM_017027869.1 | XP_016883358.1 | 8 | 1189 | |||
| XM_017027870.1 | XP_016883359.1 | 8 | 1195 | |||
| XM_017027871.1 | XP_016883360.1 | 9 | 1275 | |||
| GenBank mRNA | AF220416.1 | - | 9 | 1095 | ||
| AF258787.1 | - | 8 | 1178 | |||
| AF348207.1 | - | 7 | 1117 | |||
| AK054597.1 | - | 9 | 1449 | |||
| AK314503.1 | - | 8 | 1156 | |||
| AY027523.1 | - | 7 | 1117 | |||
| AY134747.1 | - | 8 | 1169 | |||
| BC006415.1 | - | 7 | 1052 | |||
| BC016948.1 | - | 8 | 1145 | |||
| BC048210.1 | - | 9 | 1225 | |||
| BC080598.1 | - | 8 | 1156 | |||
| BC093405.1 | - | 8 | 1192 | |||
| BC150178.1 | - | 8 | 1158 | 40148074 | ||

PHF20
Genomic Map