siRNA Details
| Gene Symbol: PHF20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016436.4 | NP_057520.2 | 10 | 1452 | ||
| XM_005260421.2 | XP_005260478.1 | 7 | 1062 | |||
| XM_011528843.1 | XP_011527145.1 | 9 | 1053 | |||
| XM_011528844.1 | XP_011527146.1 | 7 | 1124 | |||
| XM_017027864.1 | XP_016883353.1 | 11 | 1534 | |||
| XM_017027865.1 | XP_016883354.1 | 11 | 1735 | |||
| XM_017027866.1 | XP_016883355.1 | 10 | 1655 | |||
| XM_017027867.1 | XP_016883356.1 | 10 | 1490 | |||
| XM_017027868.1 | XP_016883357.1 | 8 | 1276 | |||
| XM_017027869.1 | XP_016883358.1 | 10 | 1449 | |||
| XM_017027870.1 | XP_016883359.1 | 10 | 1455 | |||
| XM_017027871.1 | XP_016883360.1 | 11 | 1535 | |||
| GenBank mRNA | AF220416.1 | - | 11 | 1355 | ||
| AF258787.1 | - | 10 | 1438 | |||
| AF348207.1 | - | 9 | 1377 | |||
| AK054597.1 | - | 11 | 1709 | |||
| AK314503.1 | - | 10 | 1416 | |||
| AY027523.1 | - | 9 | 1377 | |||
| AY134747.1 | - | 10 | 1429 | |||
| BC006415.1 | - | 9 | 1312 | |||
| BC016948.1 | - | 10 | 1405 | |||
| BC048210.1 | - | 11 | 1485 | |||
| BC080598.1 | - | 10 | 1416 | |||
| BC093405.1 | - | 10 | 1452 | |||
| BC150178.1 | - | 10 | 1418 | 40148074 | ||
| L09749.1 | - | 9 | 1332 | |||

PHF20
Genomic Map