siRNA Details
| Gene Symbol: PHF20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016436.4 | NP_057520.2 | 16 | 2535 | ||
| XM_005260421.2 | XP_005260478.1 | 13 | 2145 | |||
| XM_011528843.1 | XP_011527145.1 | 15 | 2136 | |||
| XM_011528844.1 | XP_011527146.1 | 13 | 2207 | |||
| XM_017027864.1 | XP_016883353.1 | 17 | 2617 | |||
| XM_017027865.1 | XP_016883354.1 | 17 | 2818 | |||
| XM_017027866.1 | XP_016883355.1 | 16 | 2738 | |||
| XM_017027867.1 | XP_016883356.1 | 16 | 2573 | |||
| XM_017027868.1 | XP_016883357.1 | 14 | 2359 | |||
| XM_017027869.1 | XP_016883358.1 | 16 | 2532 | |||
| XM_017027870.1 | XP_016883359.1 | 16 | 2538 | |||
| XM_017027871.1 | XP_016883360.1 | 17 | 2618 | |||
| GenBank mRNA | AK097498.1 | - | 5 | 738 | ||
| AK314503.1 | - | 16 | 2499 | |||
| AY027523.1 | - | 15 | 2460 | |||
| BC150178.1 | - | 16 | 2501 | 40148074 | ||

PHF20
Genomic Map