siRNA Details
| Gene Symbol: CRIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016441.2 | NP_057525.1 | 3 | 743 | ||
| XM_005264357.4 | XP_005264414.1 | 2 | 565 | |||
| XM_011532898.2 | XP_011531200.1 | 4 | 864 | |||
| XM_011532899.2 | XP_011531201.1 | 4 | 865 | |||
| XM_011532901.2 | XP_011531203.1 | 4 | 865 | |||
| XM_017004259.1 | XP_016859748.1 | 3 | 882 | |||
| GenBank mRNA | AB209139.1 | - | 3 | 743 | ||
| AF167706.1 | - | 3 | 716 | |||
| AY358371.1 | - | 3 | 1044 | |||
| BC111989.1 | - | 3 | 714 | 8327457 | ||
| BC113371.1 | - | 3 | 714 | 8322423 | ||

CRIM1
Genomic Map