siRNA Details
| Gene Symbol: OSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016470.7 | NP_057554.4 | 4 | 727 | ||
| XM_011528851.1 | XP_011527153.1 | 5 | 1202 | |||
| XM_011528853.2 | XP_011527155.1 | 4 | 771 | |||
| XM_011528854.2 | XP_011527156.1 | 3 | 510 | |||
| XM_017027873.1 | XP_016883362.1 | 5 | 1267 | |||
| GenBank mRNA | AF151041.1 | - | 4 | 610 | ||
| AF161517.1 | - | 4 | 610 | |||
| AF217514.1 | - | 4 | 593 | |||
| AK000476.1 | - | 4 | 631 | |||
| AK315155.1 | - | 4 | 592 | |||
| AL133000.1 | - | 4 | 612 | |||
| BC064973.1 | - | 4 | 577 | 6151501 | ||

OSER1
Genomic Map