siRNA Details
| Gene Symbol: OSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016470.7 | NP_057554.4 | 4 | 1581 | ||
| XM_011528851.1 | XP_011527153.1 | 5 | 2056 | |||
| XM_011528853.2 | XP_011527155.1 | 4 | 1625 | |||
| XM_011528854.2 | XP_011527156.1 | 3 | 1364 | |||
| XM_017027873.1 | XP_016883362.1 | 5 | 2121 | |||
| GenBank mRNA | AL133000.1 | - | 4 | 1466 | ||
| BC064973.1 | - | 4 | 1431 | 6151501 | ||

OSER1
Genomic Map