siRNA Details
| Gene Symbol: GSKIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271904.1 | NP_001258833.1 | 4 | 1013 | ||
| NM_001271905.1 | NP_001258834.1 | 4 | 566 | |||
| NM_001271906.1 | NP_001258835.1 | 3 | 498 | |||
| NM_016472.4 | NP_057556.2 | 4 | 599 | |||
| XM_017021365.1 | XP_016876854.1 | 4 | 707 | |||
| GenBank mRNA | AF151044.1 | - | 4 | 517 | ||
| AK000796.1 | - | 4 | 525 | |||
| AK094654.1 | - | 4 | 1015 | |||
| AK314423.1 | - | 4 | 525 | |||
| BC004818.2 | - | 3 | 485 | 3449501 | ||
| DA488110.1 | - | 4 | 536 | |||
| HY108584.1 | - | 3 | 474 | |||

GSKIP
Genomic Map