siRNA Details
| Gene Symbol: LY6D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003695.2 | NP_003686.1 | 3 | 351 | ||
| GenBank mRNA | AK312164.1 | - | 3 | 298 | ||
| BC022806.1 | - | 3 | 278 | |||
| BC031330.1 | - | 3 | 351 | 5428818 | ||
| BC034542.1 | - | 1 | 972 | |||
| BM019592.1 | - | 3 | 362 | |||
| X82693.1 | - | 3 | 286 | |||

LY6D
Genomic Map