siRNA Details
| Gene Symbol: GOLM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016548.3 | NP_057632.2 | 10 | 2031 | ||
| NM_177937.2 | NP_808800.1 | 10 | 2023 | |||
| GenBank mRNA | AF236056.1 | - | 10 | 1973 | ||
| AJ420432.1 | - | 1 | 254 | |||
| AK027717.1 | - | 10 | 1994 | |||
| AK075542.1 | - | 10 | 1965 | |||
| AK075557.1 | - | 10 | 1935 | |||
| AK092943.1 | - | 11 | 1911 | |||
| BC001740.1 | - | 10 | 1946 | 3351316 | ||
| EU794616.1 | - | 10 | 1990 | |||

GOLM1
Genomic Map