siRNA Details
| Gene Symbol: BFAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016561.2 | NP_057645.1 | 8 | 2228 | ||
| XM_005255350.2 | XP_005255407.1 | 7 | 2003 | |||
| XM_011522520.1 | XP_011520822.1 | 6 | 1833 | |||
| GenBank mRNA | AF173003.1 | - | 8 | 2100 | ||
| BC003054.1 | - | 8 | 2059 | 3537783 | ||

BFAR
Genomic Map