siRNA Details
| Gene Symbol: ESF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276380.1 | NP_001263309.1 | 5 | 1306 | ||
| NM_016649.3 | NP_057733.2 | 5 | 1317 | |||
| XM_017027874.1 | XP_016883363.1 | 5 | 2769 | |||
| GenBank mRNA | AF068285.1 | - | 4 | 674 | ||
| AK000375.1 | - | 5 | 1318 | |||
| AK022369.1 | - | 5 | 1268 | |||
| AK074299.1 | - | 4 | 803 | |||
| BC020952.1 | - | 4 | 936 | |||
| BC030513.1 | - | 4 | 1267 | |||
| BC046107.1 | - | 4 | 1256 | |||

ESF1
Genomic Map