siRNA Details
| Gene Symbol: ESF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276380.1 | NP_001263309.1 | 14 | 2991 | ||
| NM_016649.3 | NP_057733.2 | 14 | 3002 | |||
| XM_017027874.1 | XP_016883363.1 | 14 | 4454 | |||
| GenBank mRNA | AK022670.1 | - | 2 | 792 | ||
| AK074299.1 | - | 13 | 2488 | |||

ESF1
Genomic Map