siRNA Details
| Gene Symbol: TAS2R4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016944.1 | NP_058640.1 | 1 | 790 | ||
| GenBank mRNA | BC130439.1 | - | 1 | 837 | 40146470 | |
| BC136508.1 | - | 1 | 837 | 9020497 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TAS2R4| Gene Symbol: TAS2R4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016944.1 | NP_058640.1 | 1 | 790 | ||
| GenBank mRNA | BC130439.1 | - | 1 | 837 | 40146470 | |
| BC136508.1 | - | 1 | 837 | 9020497 | ||