siRNA Details
| Gene Symbol: KLHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257194.1 | NP_001244123.1 | 10 | 1687 | ||
| NM_001257195.1 | NP_001244124.1 | 8 | 1263 | |||
| NM_017415.2 | NP_059111.2 | 10 | 1512 | |||
| GenBank mRNA | AB032955.1 | - | 10 | 1282 | ||
| AF208068.1 | - | 10 | 1196 | |||
| AF208069.1 | - | 8 | 1300 | |||
| AF208070.1 | - | 10 | 1687 | |||
| AK300172.1 | - | 5 | 693 | |||
| AK311594.1 | - | 5 | 658 | |||
| AK314707.1 | - | 10 | 1253 | |||

KLHL3
Genomic Map