siRNA Details
| Gene Symbol: KLHL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257194.1 | NP_001244123.1 | 12 | 2027 | ||
| NM_001257195.1 | NP_001244124.1 | 10 | 1603 | |||
| NM_017415.2 | NP_059111.2 | 12 | 1852 | |||
| GenBank mRNA | AB032955.1 | - | 12 | 1622 | ||
| AF208068.1 | - | 12 | 1536 | |||
| AF208069.1 | - | 10 | 1640 | |||
| AF208070.1 | - | 12 | 2027 | |||
| AK300172.1 | - | 6 | 931 | |||
| AK311594.1 | - | 6 | 896 | |||
| AK314707.1 | - | 12 | 1593 | |||

KLHL3
Genomic Map